Learn More
Enzo Life Sciences Notch2 Recombinant monoclonal antibody (B9) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6520200
Encompass_Preferred
Clone: B9. Isotype: IgGκ. Immunogen: NRR (negative regulatory region) of mouse Notch2. Species reactivity: Human, Mouse. Specificity: Recognizes both mouse mNRR2 and human hNRR2 but not NRR1. Applications: ELISA, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3176