Learn More
Aviva Systems Biology Corp NOTCH2NLA Antibody - C-terminal region (ARP55977_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55977_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOTCH2NL. It was validated on Western Blot. . Alternate names: N2NApplicationWB. Species reactivity: Human
Catalog No. 50-286-102