Learn More
Aviva Systems Biology Corp NOTCH4 Antibody - C-terminal region (ARP38498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOTCH4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NOTCH4. Alternate names: INT3, NOTCH4ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-738