Learn More
Aviva Systems Biology Corp NOTCH4 Antibody - middle region (ARP32726_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32726_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOTCH4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NOTCH4. Alternate names: INT3, NOTCH4ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-54728