Learn More
TARGETMOL CHEMICALS INC NOTHOFAGIN 5MG
SDP
Catalog No. 502578885
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nothofagin has antioxidant and antithrombotic activities it possesses anti-inflammatory activity by inhibiting hyperpermeability expression of CAMs and adhesion and migration of leukocytes thereby endorsing its usefulness as a therapy for vascular inflammatory diseases. purity: 99%
Catalog No. 50-257-8885 Supplier TARGETMOL CHEMICALS INC Supplier No. TN19965MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nothofagin has antioxidant and antithrombotic activities it possesses anti-inflammatory activity by inhibiting hyperpermeability expression of CAMs and adhesion and migration of leukocytes thereby endorsing its usefulness as a therapy for vascular inflammatory diseases. purity: 99%