Learn More
New England Biolabs, Inc. NotI – 2500 units
SDP
Catalog No. 50812107
Encompass
A restriction endonuclease that recognizes the sequence GC^GGCC_GC.
Catalog No. 50-812-107 Supplier New England Biolabs, Inc. Supplier No. R0189L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GC^GGCC_GC.