Learn More
New England Biolabs, Inc. NotI – 500 units
SDP
Catalog No. 50812109
Encompass
A restriction endonuclease that recognizes the sequence GC^GGCC_GC.
Catalog No. 50-812-109 Supplier New England Biolabs, Inc. Supplier No. R0189S
A restriction endonuclease that recognizes the sequence GC^GGCC_GC.