Learn More
Aviva Systems Biology Corp Noto Antibody - N-terminal region (ARP36811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Noto. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of mouse Noto. Alternate names: Flh, MmNot, Not, tcApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63079