Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Notoginsenoside Fa | 88100-04-3 | MFCD29904542 | 99.0% | 1241.41 | C59H100O27 | 10 MG
SDP

Catalog No. 5000288941
Encompass_Preferred

Notoginsenoside Fa is a protopanaxadiol (PPD)-type saponin isolated from Panax notoginseng. Supplied as a white to off-white solid for research use, it has been studied for potential neuroprotective activity, including activation and recovery of degenerated brain function.

  • Purity: 99.0%
  • Cas number: 88100-04-3
  • Chemical formula: C59H100O27
  • Molecular weight: 1241.41
  • Appearance: white to off-white solid
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-88941 Supplier Medchemexpress LLC Supplier No. HYN253010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Notoginsenoside Fa is a protopanaxadiol (PPD)-type saponin isolated from Panax notoginseng. Supplied as a white to off-white solid for research use, it has been studied for potential neuroprotective activity, including activation and recovery of degenerated brain function.

  • Purity: 99.0%
  • Cas number: 88100-04-3
  • Chemical formula: C59H100O27
  • Molecular weight: 1241.41
  • Appearance: white to off-white solid
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.