Learn More
Medchemexpress LLC Notoginsenoside Fc | 88122-52-5 | 5 MG
SDP
Catalog No. 5000421512
Encompass_Preferred
Notoginsenoside Fc is a protopanaxadiol- (PPD-) type saponin isolated from the leaves of Panax notoginseng. It effectively counteracts platelet aggregation and can accelerate reendothelialization following vascular injury in diabetic rats by promoting autophagy.
  • Effectively counteracts platelet aggregation
  • Accelerates reendothelialization following vascular injury in diabetic rats by promoting autophagy
  • Markedly upregulates the expression of LC3B and Beclin 1, and downregulates that of p62 in RAOECs (in vitro)
  • Alleviates excessive neointimal formation following carotid artery injury in diabetic Sprague-Dawley rats (in vivo)
Catalog No. 50-004-21512 Supplier Medchemexpress LLC Supplier No. HYN25315MG
Notoginsenoside Fc is a protopanaxadiol- (PPD-) type saponin isolated from the leaves of Panax notoginseng. It effectively counteracts platelet aggregation and can accelerate reendothelialization following vascular injury in diabetic rats by promoting autophagy.
  • Effectively counteracts platelet aggregation
  • Accelerates reendothelialization following vascular injury in diabetic rats by promoting autophagy
  • Markedly upregulates the expression of LC3B and Beclin 1, and downregulates that of p62 in RAOECs (in vitro)
  • Alleviates excessive neointimal formation following carotid artery injury in diabetic Sprague-Dawley rats (in vivo)