Learn More
Cayman Chemical Notoginsenoside Fe, Purity: ≥90%, CAS: 88105-29-7, 25 mg
SDP
Catalog No. 502948371
Encompass_Preferred
A saponin and metabolite of ginsenoside Rc; cytotoxic to L1210 cells (IC50 = 80 µM)
Catalog No. 50-294-8371 Supplier Cayman Chemical Supplier No. 3429725 MG
A saponin and metabolite of ginsenoside Rc; cytotoxic to L1210 cells (IC50 = 80 µM)