Learn More
Medchemexpress LLC Notoginsenoside R1 250mg
SDP
Catalog No. 5000669399
Encompass_Preferred
Notoginsenoside R1 (Sanchinoside R1) a saponin is isolated from P notoginseng Notoginsenoside R1 exhibits anti-oxidation anti-inflammatory anti-angiogenic and anti-apoptosis activities Notoginsenoside R1 provides cardioprotection against ischemia/reperfusion (I/R) injury Notoginsenoside R1 also provides neuroprotection in H2O2-induced oxidative damage in PC12 cells[1][2][3]
Catalog No. 50-006-69399 Supplier Medchemexpress LLC Supplier No. 250MGHYN0615
Add to Cart
Add to Cart
Notoginsenoside R1 (Sanchinoside R1) a saponin is isolated from P notoginseng Notoginsenoside R1 exhibits anti-oxidation anti-inflammatory anti-angiogenic and anti-apoptosis activities Notoginsenoside R1 provides cardioprotection against ischemia/reperfusion (I/R) injury Notoginsenoside R1 also provides neuroprotection in H2O2-induced oxidative damage in PC12 cells[1][2][3]