Learn More
Cayman Chemical Notoginsenoside R1, Purity: ≥98%, CAS: 80418-24-2, 50 mg
SDP
Catalog No. 502991266
Encompass_Preferred
A saponin; decreases serum total cholesterol, triacylglycerol, and oxLDL levels, as well as reduces atherosclerotic lesion area and lipid deposition in the aortic root of ApoE-/- mice fed a Western diet at 25 mg/kg per day for 8 weeks; inhibits apoptosis of hippocampal neurons and reduces infarct size in a rat model of cerebral ischemia-reperfusion injury induced by bilateral common carotid artery occlusion at 100 mg/kg; decreases colonic NF-ĸB and MPO activities and TNF-α and IL-6 levels, as well as reduces colonic shortening, tissue damage, and inflammatory cell infiltration in a DSS-induced mouse model of colitis at 25 mg/kg per day for 7 days
Catalog No. 50-299-1266 Supplier Cayman Chemical Supplier No. 2969150 MG
A saponin; decreases serum total cholesterol, triacylglycerol, and oxLDL levels, as well as reduces atherosclerotic lesion area and lipid deposition in the aortic root of ApoE-/- mice fed a Western diet at 25 mg/kg per day for 8 weeks; inhibits apoptosis of hippocampal neurons and reduces infarct size in a rat model of cerebral ischemia-reperfusion injury induced by bilateral common carotid artery occlusion at 100 mg/kg; decreases colonic NF-ĸB and MPO activities and TNF-α and IL-6 levels, as well as reduces colonic shortening, tissue damage, and inflammatory cell infiltration in a DSS-induced mouse model of colitis at 25 mg/kg per day for 7 days