Learn More
StatLab NOTOX Histo Formalin-Free Fixative

Description
- Non-carcinogenic, non-noxious, non-corrosive, heavy metals and chloride ion free fixative
- Fix all sizes of specimens from small sections to whole organs
- Supports immunohistochemical staining, in-situ hybridization, and PCR techniques
- No pre-mixing or dilution
- Available in 1 gallon or 5 gallon packages
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Histological |
| Product Type | Fixative |
| Quantity | 1 gal. |
| Solution Type | Fixative |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.