Learn More
Description
- Pre-printed message mats warn employees or customers who may be entering a hazardous area
- Offer functionality as an entrance mat, cleaning dirt and moisture from shoes while keeping facilities cleaner and safer.
- Safety Message Mats should be used indoors at entrances, lobbies, foyers, or any other light to heavy traffic areas where a safety message should be seen.
- Highly visible colors and graphics allow for immediate identification
- 24 ounce nylon top surface provides excellent moisture absorption and retention
- Vinyl backing ensure minimal buckling and mat movement
Specifications
Specifications
| Material | Nylon, Vinyl |
| Dimensions (L x W) | 5 x 3 ft. |
| Color | Blue/White |
| Type | Entrance Mat |
| Thickness (English) | 0.38 in. |
| Thickness (Metric) | 0.97 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.