Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notrax™ 194GS Germs Stop Here Mat

Catalog No. 19085901 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
5 x 3 ft.
6 x 4 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-085-901 5 x 3 ft.
19-085-902 6 x 4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-085-901 Supplier NoTrax Supplier No. 194SGS35BU

Safety Message mats make safety message loud and clear while keeping facilities cleaner and safer.

  • Pre-printed message mats warn employees or customers who may be entering a hazardous area
  • Offer functionality as an entrance mat, cleaning dirt and moisture from shoes while keeping facilities cleaner and safer.
  • Safety Message Mats should be used indoors at entrances, lobbies, foyers, or any other light to heavy traffic areas where a safety message should be seen.
  • Highly visible colors and graphics allow for immediate identification
  • 24 ounce nylon top surface provides excellent moisture absorption and retention
  • Vinyl backing ensure minimal buckling and mat movement

Specifications

Dimensions (L x W) 5 x 3 ft.
Material Nylon, Vinyl
Thickness (Metric) 0.97 cm
Color Blue/White
Type Entrance Mat
Thickness (English) 0.38 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.