Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Notrax™ 448 Autoclavable Bubble Antimicrobial Mat

Catalog No. 19111107 Shop All Justrite Safety Group Products
Change view
Click to view available options
Dimensions (L x W):
3 x 2 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-111-107 3 x 2 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-111-107 Supplier Justrite™ Supplier No. 448S0023GY
This item is not returnable. View return policy

Notrax™ 448 Autoclavable Bubble Antimicrobial Mat is a rubber anti-fatigue floor mat with a bubble top design. Offset bubbles on both the top and bottom of the mat provides flex and creates airflow resulting in maximum comfort for workers required to be on their feet for long periods of time.

  • Industrial grade antimicrobial agent that inhibits the growth of most Gram positive and Gram-negative bacteria and fungus is manufactured directly into the rubber compound, ensuring that the antimicrobial properties won't wear-off over time like many post applied topical agents.
  • Intended for use in dry environments, but easy to wipe clean
  • Durable rubber compound is highly resistant to many hospital grade cleaners and disinfectants so occasional spills can be cleaned up quickly and easily.
  • Molded beveled edges on all four sides eliminates trip hazards by allowing for easy access onto and off the mat.
  • Ideal Environments: Sterile environments such as labs, hospitals, surgery centers, or pharmaceutical.
  • Active ingredient is a broad spectrum anti-microbial agent mostly anti-bacterial with limited anti-fungal and anti-viral properties
  • Withstands the harsh conditions of the sterilization process through use of chemicals or an autoclave
  • Bubbles are molded on an offset on the top and bottom of the mat to maximize flex and create air flow, all to enhance the ergonomic benefit
  • Prevents contamination in sanitized commercial and industrial sites
  • Beveled edges on all 4 sides

Specifications

Width (Metric) 61 cm
Thickness (Metric) 0.97 cm
Length (English) 3 ft.
Color Gray
Description Lightweight mat, easy to move to clean the floor underneath
Dimensions (L x W) 3 x 2 ft.
Width (English) 2 ft.
Length (Metric) 91.4 cm
Thickness (English) 0.38 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.