Learn More
MSC NoTrax Entrance Mat, Prelude, Non Absorbent Scraper, Indoor/Outdoor, 1 ea
Catalog No. 5000793911
Encompass
Prelude™ is a scraper mat composed of coarse, loop-twisted non-absorbent fibers, designed for placement outside entrances to effectively remove dirt and debris from foot traffic. Its aggressive fibers resist crushing while trapping debris and preventing it from entering the facility. This mat can function as a standalone mat or as the initial stage of the coordinated 'Stages' entrance matting system.
  • Coarse loop-twisted non-absorbent fiber to scrape heavy debris from foot traffic
  • Aggressive fibers resist crushing while entrapping debris and preventing it from entering the facility
  • Low profile entrance mat to perform in narrow clearance doorways
  • Vinyl backing ensures minimal buckling and mat movement
  • Can be used as the first section (scraping) of the 'Stages' entrance mat system
Catalog No. 50-007-93911 Supplier MSC Supplier No. 71217731
Prelude™ is a scraper mat composed of coarse, loop-twisted non-absorbent fibers, designed for placement outside entrances to effectively remove dirt and debris from foot traffic. Its aggressive fibers resist crushing while trapping debris and preventing it from entering the facility. This mat can function as a standalone mat or as the initial stage of the coordinated 'Stages' entrance matting system.
  • Coarse loop-twisted non-absorbent fiber to scrape heavy debris from foot traffic
  • Aggressive fibers resist crushing while entrapping debris and preventing it from entering the facility
  • Low profile entrance mat to perform in narrow clearance doorways
  • Vinyl backing ensures minimal buckling and mat movement
  • Can be used as the first section (scraping) of the 'Stages' entrance mat system