Learn More
Cayman Chemical notuberostemonIn 10mg
SDP
Catalog No. 502989468
Encompass_Preferred
An alkaloid; inhibits LPS-induced increases in iNOS protein levels and NO production in RAW 264.7 cells at 100 µM; inhibits RANKL-induced osteoclast differentiation of RAW 264.7 cells at 50 µM; reduces the citric acid-induced cough reflex in conscious guinea pigs at 133 µmol/kg; inhibits pulmonary collagen deposition and fibrosis, as well as BALF monocyte and lymphocyte infiltration in a mouse model of bleomycin-induced pulmonary fibrosis at 40 mg/kg
Catalog No. 50-298-9468 Supplier Cayman Chemical Supplier No. 3011610 MG
An alkaloid; inhibits LPS-induced increases in iNOS protein levels and NO production in RAW 264.7 cells at 100 µM; inhibits RANKL-induced osteoclast differentiation of RAW 264.7 cells at 50 µM; reduces the citric acid-induced cough reflex in conscious guinea pigs at 133 µmol/kg; inhibits pulmonary collagen deposition and fibrosis, as well as BALF monocyte and lymphocyte infiltration in a mouse model of bleomycin-induced pulmonary fibrosis at 40 mg/kg