Learn More
Cayman Chemical Nourseothricin (sulfate), 96736-11-7, 1 mg
SDP
Supplier:  Cayman Chemical 162271
Encompass_Preferred
A broad-spectrum antibiotic that inhibits protein synthesis by inducing miscoding; commonly used as a dominant selection antibiotic for genetically modified bacteria, yeasts, fungi, protozoa, plants, and mammalian cells
Catalog No. NC1267339