Learn More
Gold Biotechnology Inc Nourseothricin Sulfate (Streptothricin Sulfate) (NTC or clonNAT) 5 x 1 g
SDP
Catalog No. NC1755445
Encompass
Nourseothricin is a broad-spectrum antibiotic derived from Streptomyces noursei. In addition to being effective on prokaryotic cells of gram-positive and gram-negative bacteria, various fungi including Candida albicans, and certain DNA and RNA viruses, it is also effective on eukaryotic cells of higher eukaryotes. It has been used as a selection agent for cells containing the gene conferring resistance to it.
Catalog No. NC1755445 Supplier Gold Biotechnology Inc Supplier No. N5005
Nourseothricin is a broad-spectrum antibiotic derived from Streptomyces noursei. In addition to being effective on prokaryotic cells of gram-positive and gram-negative bacteria, various fungi including Candida albicans, and certain DNA and RNA viruses, it is also effective on eukaryotic cells of higher eukaryotes. It has been used as a selection agent for cells containing the gene conferring resistance to it.