Learn More
TARGETMOL CHEMICALS INC NOURSEOTHRICIN SULFATE 50MG
SDP
Catalog No. 502581476
Encompass_Preferred
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. Nourseothricin sulfate (Streptothricin sulfate) is a broad-spectrum antibioticand inhibits protein biosynthesis in prokaryotic cells and strongly inhibits the growth of eukaryotes like fungi . purity: 100%
Catalog No. 50-258-1476 Supplier TARGETMOL CHEMICALS INC Supplier No. T1224850MG
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. Nourseothricin sulfate (Streptothricin sulfate) is a broad-spectrum antibioticand inhibits protein biosynthesis in prokaryotic cells and strongly inhibits the growth of eukaryotes like fungi . purity: 100%