Learn More
Medchemexpress LLC HY-129065 5mg Medchemexpress, Nourseothricin (sulfate) CAS:96736-11-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1290655MG
Encompass_Preferred
Medchemexpress, HY-129065 5mg Nourseothricin (sulfate) CAS:96736-11-7 Nourseothricin sulfate (Streptothricin sulfate) is a broad-spectrum antibiotic that destroys the outer membrane of Gram-negative bacteria and is a dominant selective marker for Fonsecaea pedrosoi[2]. Nourseothricin sulfate inhibits protein biosynthesis in prokaryotic cells and strongly inhibits the growth of eukaryotes like fungi and can also be used as a elective marker for a wide range of organisms including bacteria, yeast, filamentous fungi, and plant cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0150