Learn More
Gold Biotechnology Inc Nourseothricin Sulfate (Streptothricin Sulfate) (NTC or clonNAT) 1 g
SDP
Catalog No. 501532817
Encompass_Preferred
Nourseothricin is a broad-spectrum antibiotic derived from Streptomyces noursei. In addition to being effective on prokaryotic cells of gram-positive and gram-negative bacteria, various fungi including Candida albicans, and certain DNA and RNA viruses, it is also effective on eukaryotic cells of higher eukaryotes. It has been used as a selection agent for cells containing the gene conferring resistance to it.
Catalog No. 50-153-2817 Supplier Gold Biotechnology Inc Supplier No. N5001
Nourseothricin is a broad-spectrum antibiotic derived from Streptomyces noursei. In addition to being effective on prokaryotic cells of gram-positive and gram-negative bacteria, various fungi including Candida albicans, and certain DNA and RNA viruses, it is also effective on eukaryotic cells of higher eukaryotes. It has been used as a selection agent for cells containing the gene conferring resistance to it.