Learn More
Apexbio Technology LLC Nourseothricin (sulfate) 96736-11-7 10mg
SDP
Catalog No. 5000091433
Encompass_Preferred
Nourseothricin sulfate (CAS 96736-11-7) is a broad-spectrum antibiotic composed primarily of streptothricins D and F It exerts its biological activity by penetrating bacterial outer membranes via porin channels and inducing membrane permeability alterations thereby disrupting cellular integrity and inhibiting protein biosynthesis in prokaryotes In experimental studies nourseothricin sulfate is used as an antibiotic selection reagent particularly for selecting transformed cells expressing nourseothricin resistance genes Its IC50 values depend significantly on bacterial strains and assay conditions Based on these pharmacological properties nourseothricin sulfate holds research potential as a selective marker in molecular biology for bacteria yeast filamentous fungi and plant cells
Catalog No. 50-000-91433 Supplier Apexbio Technology LLC Supplier No. C515510
Nourseothricin sulfate (CAS 96736-11-7) is a broad-spectrum antibiotic composed primarily of streptothricins D and F It exerts its biological activity by penetrating bacterial outer membranes via porin channels and inducing membrane permeability alterations thereby disrupting cellular integrity and inhibiting protein biosynthesis in prokaryotes In experimental studies nourseothricin sulfate is used as an antibiotic selection reagent particularly for selecting transformed cells expressing nourseothricin resistance genes Its IC50 values depend significantly on bacterial strains and assay conditions Based on these pharmacological properties nourseothricin sulfate holds research potential as a selective marker in molecular biology for bacteria yeast filamentous fungi and plant cells