Learn More
Medchemexpress LLC Nourseothricin (sulfate) | 96736-11-7 | 87.3% | 50 MG
SDP
Catalog No. 5000340000
Encompass_Preferred
Nourseothricin sulfate is a broad-spectrum antibiotic that targets the outer membrane of Gram-negative bacteria. It inhibits protein biosynthesis in prokaryotic cells and significantly hinders the growth of eukaryotes like fungi. This antibiotic can also serve as a selective marker for a diverse range of organisms.
  • Broad-spectrum antibiotic activity
  • Destroys the outer membrane of Gram-negative bacteria
  • Inhibits protein biosynthesis in prokaryotic cells
  • Strongly inhibits the growth of eukaryotes such as fungi
  • Useful as an elective marker for various organisms, including bacteria, yeast, filamentous fungi, and plant cells
Catalog No. 50-003-40000 Supplier Medchemexpress LLC Supplier No. HY12906550MG
Nourseothricin sulfate is a broad-spectrum antibiotic that targets the outer membrane of Gram-negative bacteria. It inhibits protein biosynthesis in prokaryotic cells and significantly hinders the growth of eukaryotes like fungi. This antibiotic can also serve as a selective marker for a diverse range of organisms.
  • Broad-spectrum antibiotic activity
  • Destroys the outer membrane of Gram-negative bacteria
  • Inhibits protein biosynthesis in prokaryotic cells
  • Strongly inhibits the growth of eukaryotes such as fungi
  • Useful as an elective marker for various organisms, including bacteria, yeast, filamentous fungi, and plant cells