Learn More
Medchemexpress LLC Nourseothricin (sulfate) | 96736-11-7 | 99.9% | 1 G
SDP
Catalog No. 5000339380
Encompass_Preferred
Nourseothricin sulfate is a broad-spectrum antibiotic that inhibits protein biosynthesis in prokaryotic cells and destroys the outer membrane of Gram-negative bacteria. It also functions as a selective marker for *Fonsecaea pedrosoi* and is effective for a diverse range of organisms including bacteria, yeast, filamentous fungi, and plant cells.
  • Broad-spectrum antibiotic
  • Destroys outer membrane of Gram-negative bacteria
  • Inhibits protein biosynthesis in prokaryotic cells
  • Inhibits growth of eukaryotes such as fungi
  • Selective marker for *Fonsecaea pedrosoi*
  • Used as an elective marker for various organisms
Catalog No. 50-003-39380 Supplier Medchemexpress LLC Supplier No. HY1290651G
Nourseothricin sulfate is a broad-spectrum antibiotic that inhibits protein biosynthesis in prokaryotic cells and destroys the outer membrane of Gram-negative bacteria. It also functions as a selective marker for *Fonsecaea pedrosoi* and is effective for a diverse range of organisms including bacteria, yeast, filamentous fungi, and plant cells.
  • Broad-spectrum antibiotic
  • Destroys outer membrane of Gram-negative bacteria
  • Inhibits protein biosynthesis in prokaryotic cells
  • Inhibits growth of eukaryotes such as fungi
  • Selective marker for *Fonsecaea pedrosoi*
  • Used as an elective marker for various organisms