Learn More
Medchemexpress LLC Nourseothricin (sulfate) | 96736-11-7 | 99.9% | 10 MG
SDP
Catalog No. 5000341321
Encompass_Preferred
Nourseothricin sulfate is a broad-spectrum antibiotic that inhibits protein biosynthesis in prokaryotic cells and strongly inhibits the growth of eukaryotes such as fungi. It destroys the outer membrane of Gram-negative bacteria and acts as a dominant selective marker for Fonsecaea pedrosoi, also serving as a selective marker for various other organisms.
  • Broad-spectrum antibiotic
  • Destroys outer membrane of Gram-negative bacteria
  • Acts as a dominant selective marker for Fonsecaea pedrosoi
  • Inhibits protein biosynthesis in prokaryotic cells
  • Strongly inhibits the growth of eukaryotes such as fungi
  • Selective marker for bacteria, yeast, filamentous fungi, and plant cells
Catalog No. 50-003-41321 Supplier Medchemexpress LLC Supplier No. HY12906510MG
Nourseothricin sulfate is a broad-spectrum antibiotic that inhibits protein biosynthesis in prokaryotic cells and strongly inhibits the growth of eukaryotes such as fungi. It destroys the outer membrane of Gram-negative bacteria and acts as a dominant selective marker for Fonsecaea pedrosoi, also serving as a selective marker for various other organisms.
  • Broad-spectrum antibiotic
  • Destroys outer membrane of Gram-negative bacteria
  • Acts as a dominant selective marker for Fonsecaea pedrosoi
  • Inhibits protein biosynthesis in prokaryotic cells
  • Strongly inhibits the growth of eukaryotes such as fungi
  • Selective marker for bacteria, yeast, filamentous fungi, and plant cells