Learn More
Medchemexpress LLC Nourseothricin sulfate | 96736-11-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY129065
Encompass_Preferred
Nourseothricin sulfate is a broad-spectrum antibiotic that targets the outer membrane of Gram-negative bacteria. It inhibits protein biosynthesis in prokaryotic cells and suppresses the growth of eukaryotes like fungi. It can also be utilized as a selective marker for a wide array of organisms.
  • Broad-spectrum antibiotic
  • Targets outer membrane of Gram-negative bacteria
  • Serves as a dominant selective marker for *Fonsecaea pedrosoi*
  • Inhibits protein biosynthesis in prokaryotic cells
  • Suppresses the growth of eukaryotes like fungi
  • Can be used as a selective marker for various organisms, including bacteria, yeast, filamentous fungi, and plant cells
Catalog No. 50-003-42210