Learn More
Sigma Aldrich Fine Chemicals Biosciences Nourseothricin sulfate sui50MG
Catalog No. 502907132
Encompass
Chemical structure peptidyl nucleoside aminoglycoside antibiotic. Nourseothricin (NTC) is a mixture of the related compounds streptothricin C D E and F.
Catalog No. 50-290-7132 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N018650MG
Chemical structure peptidyl nucleoside aminoglycoside antibiotic. Nourseothricin (NTC) is a mixture of the related compounds streptothricin C D E and F.