Learn More
Cayman Chemical NourseothrIcInsulfate 50mg
SDP
Catalog No. 502972345
Encompass_Preferred
A broad-spectrum antibiotic that inhibits protein synthesis by inducing miscoding; commonly used as a dominant selection antibiotic for genetically modified bacteria, yeasts, fungi, protozoa, plants, and mammalian cells
Catalog No. 50-297-2345 Supplier Cayman Chemical Supplier No. 1622750 MG
A broad-spectrum antibiotic that inhibits protein synthesis by inducing miscoding; commonly used as a dominant selection antibiotic for genetically modified bacteria, yeasts, fungi, protozoa, plants, and mammalian cells