Learn More
Aviva Systems Biology Corp NOV Antibody - C-terminal region (ARP43613_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43613_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NOV. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NOV. Alternate names: CCN3, IGFBP9, NOVHApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-58588