Learn More
Aviva Systems Biology Corp NOV Antibody - middle region (ARP43612_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43612_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NOV. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NOV. Alternate names: CCN3, IGFBP9, NOVHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58587