Learn More
Medchemexpress LLC Nov/CCN3 protein, human (recombinant, Sf9, C-His tag) | 97.1% | 20UG
SDP
Catalog No. 5000174884
Encompass_Preferred
Recombinant human NOV/CCN3 protein produced in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in Tris/NaCl buffer with glycerol; suitable for biochemical assays and cell signaling studies. Analytical testing indicates ~97.1% purity by reducing SDS-PAGE, an observed molecular weight of approximately 47 kDa, and endotoxin <1 EU/μg.
  • Recombinant human protein expressed in Sf9 insect cells.
  • C-terminal His tag for affinity purification and detection.
  • High purity (~97.1%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Supplied in buffered, glycerol-containing solution for stability.
Catalog No. 50-001-74884 Supplier Medchemexpress LLC Supplier No. HYP70132720UG
Recombinant human NOV/CCN3 protein produced in Sf9 insect cells with a C-terminal His tag. Supplied as a 0.22 μm filtered solution in Tris/NaCl buffer with glycerol; suitable for biochemical assays and cell signaling studies. Analytical testing indicates ~97.1% purity by reducing SDS-PAGE, an observed molecular weight of approximately 47 kDa, and endotoxin <1 EU/μg.
  • Recombinant human protein expressed in Sf9 insect cells.
  • C-terminal His tag for affinity purification and detection.
  • High purity (~97.1%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Supplied in buffered, glycerol-containing solution for stability.