Learn More
BURTON MEDICAL LLC NOVA EXAM LIGHT FLOORSTND 120V
Catalog No. NC0645049
Encompass_Preferred
25,000 lux (low level) (2323 fc) at 18" (46 cm). 50, 000 lux (high level) (4645 fc) at 18" (46 cm). 4600 K color remperature. CRI of 93. 40, 000 hour average LED life. Floor Stand w/ casters.
Catalog No. NC0645049 Supplier BURTON MEDICAL LLC Supplier No. NXFL
25,000 lux (low level) (2323 fc) at 18" (46 cm). 50, 000 lux (high level) (4645 fc) at 18" (46 cm). 4600 K color remperature. CRI of 93. 40, 000 hour average LED life. Floor Stand w/ casters.