Learn More
Electron Microscopy Sciences Oncyte Slide Nova, 64 well, 2.5 x 2.5 mm I.D.
SDP
Catalog No. 5027536
Encompass_Preferred
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. NOVA for highest fluorescent sensitivity but reduced binding capacity.
Wells: 64
I.D.: 2.5 x 2.5 mm
Catalog No. 50-275-36 Supplier Electron Microscopy Sciences Supplier No. 7033558N
ONCYTE is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding. It also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. NOVA for highest fluorescent sensitivity but reduced binding capacity.
Wells: 64
I.D.: 2.5 x 2.5 mm