Learn More
Waters Corp Waters Corporation Column, C18, 4 μm, 3.9 mm x 75 mm
Supplier:  Waters Corp WAT011670
Encompass_Preferred
Nova-Pak C18 columns are general-purpose, silica-based, reversed-phase C18 columns utilizing 4 μm particle technology. They deliver higher resolution, more efficient, and faster chromatography than 5 μm columns, allowing shorter column lengths and reduced solvent consumption. These columns can also resolve complex mixtures when combined with longer columns. Manufactured under strict cGMP and ISO 9002 guidelines, they ensure high precision and consistent, repeatable results.
  • Provides high resolution and efficient separations.
  • Reduces solvent consumption by enabling shorter column lengths.
  • Suitable for complex and challenging separations.
  • Offers high precision and a wide range of separation capabilities.
  • Lowers operating expenses through quicker separations.
  • Ensures batch-to-batch reproducibility, high column efficiency, peak symmetry, and extended column lifetime.
Catalog No. 50-785-042