Learn More
Waters Corp Column, C18, 60 Å, 4 μm, 4.6 mm X 150 mm
Catalog No. 502392470 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 columns are general-purpose, silica-based, reversed-phase columns that use 4 μm particle technology. They offer higher resolution, more efficient separations, and faster chromatography compared to 5 μm columns. Each column includes an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history. While compatible with any HPLC system, full advantages are fully realized with the Waters Alliance iS System running Empower software.
  • C18 chemistry for reversed-phase separation
  • Silica particle substrate
  • 60 Å pore size
  • 4 μm particle size
  • 4.6 mm inner diameter and 150 mm length
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • Electronic eConnect column tag for automatic identification and tracking
Catalog No. 50-239-2470 Supplier Waters Corp Supplier No. WAT044375RF
These C18 columns are general-purpose, silica-based, reversed-phase columns that use 4 μm particle technology. They offer higher resolution, more efficient separations, and faster chromatography compared to 5 μm columns. Each column includes an electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history. While compatible with any HPLC system, full advantages are fully realized with the Waters Alliance iS System running Empower software.
  • C18 chemistry for reversed-phase separation
  • Silica particle substrate
  • 60 Å pore size
  • 4 μm particle size
  • 4.6 mm inner diameter and 150 mm length
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • Electronic eConnect column tag for automatic identification and tracking