Learn More
Waters Corp Nova-Pak C8 Radial-Pak Column, 60Å, 4 µm, 5 mm X 100 mm, 1/pk
Catalog No. 50785379 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak C8 columns are general purpose, silica-based, reversed-phase C8 columns that are based on 4 µm particle technology. Compared to a 5 µm column, Nova-Pak columns offer higher resolution, more efficient separations, and faster chromatography.
Catalog No. 50-785-379 Supplier Waters Corp Supplier No. WAT035890
Nova-Pak C8 columns are general purpose, silica-based, reversed-phase C8 columns that are based on 4 µm particle technology. Compared to a 5 µm column, Nova-Pak columns offer higher resolution, more efficient separations, and faster chromatography.