Learn More
Waters Corp WATERS CORPORATION Column, Nova-Pak C8, 60Å, 4 μm, 3.9 mm X 150 mm
Catalog No. 50785375 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak C8 columns are general-purpose, silica-based, reversed-phase C8 columns utilizing 4 μm particle technology. They offer higher resolution, efficient separations, and faster chromatography compared to 5 μm columns. These columns reduce solvent consumption and maintain resolution for complex mixtures, enhancing lab efficiency. Suitable for various system configurations and instrument platforms, they withstand pressures up to 6000 psi (415 Bar).
  • Higher resolution and efficient separations.
  • Faster chromatography, reducing wait times.
  • Improved separation via larger surface area for mobile phase interaction.
  • Reduced solvent consumption without compromising resolution.
  • Compatible with diverse system configurations and instrument platforms.
  • Equipped with end caps for optimal security and ease of use.
  • Strict quality control for high batch-to-batch reproducibility.
Catalog No. 50-785-375 Supplier Waters Corp Supplier No. WAT035876
Add to Cart
Add to Cart
Nova-Pak C8 columns are general-purpose, silica-based, reversed-phase C8 columns utilizing 4 μm particle technology. They offer higher resolution, efficient separations, and faster chromatography compared to 5 μm columns. These columns reduce solvent consumption and maintain resolution for complex mixtures, enhancing lab efficiency. Suitable for various system configurations and instrument platforms, they withstand pressures up to 6000 psi (415 Bar).
  • Higher resolution and efficient separations.
  • Faster chromatography, reducing wait times.
  • Improved separation via larger surface area for mobile phase interaction.
  • Reduced solvent consumption without compromising resolution.
  • Compatible with diverse system configurations and instrument platforms.
  • Equipped with end caps for optimal security and ease of use.
  • Strict quality control for high batch-to-batch reproducibility.