Learn More
Waters Corp Waters Column, Cyano (CN), 4 μm, 60 Å, 3.9 mm × 300 mm
Supplier:  Waters Corp WAT056920
Encompass_Preferred
These silica-based, cyano phase columns utilize 4 μm particle technology, offering unique compound selectivity for normal-phase and reversed-phase applications. The small 4 μm particle size provides higher resolution and faster chromatography, enabling accurate results and a wide range of separation possibilities.
  • Achieves higher resolution and faster chromatography with 4 μm particle technology.
  • Reduces solvent consumption through shorter column lengths, or resolves complex mixtures with longer columns.
  • Delivers accurate and consistent results due to high-efficiency particles.
  • Lowers lab operating costs by enabling faster separations and reduced solvent usage.
  • Manufactured under strict cGMP, ISO 9002 conditions for high quality.
  • Features excellent column efficiency, peak symmetry, lifetime, and reproducibility.
  • Operates effectively across a pH range of 2–8.
Catalog No. 50-785-831