Learn More
Waters Corp Nova-Pak Cyano (CN) Column, 60Å, 4 µm, 3.9 mm X 75 mm, 1/pk
Catalog No. 50784970 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak HP CN columns are silica-based, cyano phases that are based on 4 µm particle technology. These columns can be used to achieve unique compound selectivity for both normal-phase and reversed-phase applications.
Catalog No. 50-784-970 Supplier Waters Corp Supplier No. WAT010270
Nova-Pak HP CN columns are silica-based, cyano phases that are based on 4 µm particle technology. These columns can be used to achieve unique compound selectivity for both normal-phase and reversed-phase applications.