Learn More
Waters Corp Waters Nova-Pak Cyano Guard Cartridge, 4 μm, 3.9 mm × 20 mm
Catalog No. 50785625 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Nova-Pak HP CN guard cartridges are silica-based, cyano phases with 4 μm particle technology, offering unique selectivity for normal and reversed-phase applications. They protect HPLC cartridges, ensuring accurate results by retaining polar compounds, and seamlessly integrate with various HPLC solvents. End-capped to reduce column bleed and interference, they enhance separation efficiency and provide clean peaks.
  • Silica-based, cyano phases, 4 μm particle.
  • Unique selectivity for normal and reversed-phase.
  • Protects HPLC cartridges from contamination.
  • Ensures accurate results, retains polar compounds.
  • Compatible with various HPLC solvents.
  • End-capped design reduces column bleed/interference.
  • Enhances separation efficiency with clean peaks.
  • Extends HPLC cartridge lifetime.
  • Suitable for pharmaceuticals, food, environmental samples.
Catalog No. 50-785-625 Supplier Waters Corp Supplier No. WAT046840
These Nova-Pak HP CN guard cartridges are silica-based, cyano phases with 4 μm particle technology, offering unique selectivity for normal and reversed-phase applications. They protect HPLC cartridges, ensuring accurate results by retaining polar compounds, and seamlessly integrate with various HPLC solvents. End-capped to reduce column bleed and interference, they enhance separation efficiency and provide clean peaks.
  • Silica-based, cyano phases, 4 μm particle.
  • Unique selectivity for normal and reversed-phase.
  • Protects HPLC cartridges from contamination.
  • Ensures accurate results, retains polar compounds.
  • Compatible with various HPLC solvents.
  • End-capped design reduces column bleed/interference.
  • Enhances separation efficiency with clean peaks.
  • Extends HPLC cartridge lifetime.
  • Suitable for pharmaceuticals, food, environmental samples.