Learn More
Waters Corp Waters Nova-Pak Phenyl Sentry Guard Cartridge, Phenyl, 4 μm, 3.9 mm x 20 mm
Catalog No. 50785624 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge extends HPLC column life by reducing particle matter and chemical pollutants. It is compatible with 4 μm silica-based particle columns, utilizing a phenyl ligand for unique compound selectivity in reversed-phase applications. It traps particulates and strongly retained compounds, preventing contamination and ensuring consistent results.
  • Extends HPLC column life by reducing particle matter and chemical pollutants.
  • Compatible with 4 μm silica-based particle columns.
  • Provides unique compound selectivity in reversed-phase applications.
  • Features two holder designs for tool-free swapping.
  • Supports a pH range of 2 to 8.
  • Operates at a maximum pressure of 6000 psi (415 Bar).
  • Equipped with 4 μm particle size and 60 Å pore size.
  • Offers enhanced selectivity for aromatic compounds.
Catalog No. 50-785-624 Supplier Waters Corp Supplier No. WAT046835
This Sentry Guard Cartridge extends HPLC column life by reducing particle matter and chemical pollutants. It is compatible with 4 μm silica-based particle columns, utilizing a phenyl ligand for unique compound selectivity in reversed-phase applications. It traps particulates and strongly retained compounds, preventing contamination and ensuring consistent results.
  • Extends HPLC column life by reducing particle matter and chemical pollutants.
  • Compatible with 4 μm silica-based particle columns.
  • Provides unique compound selectivity in reversed-phase applications.
  • Features two holder designs for tool-free swapping.
  • Supports a pH range of 2 to 8.
  • Operates at a maximum pressure of 6000 psi (415 Bar).
  • Equipped with 4 μm particle size and 60 Å pore size.
  • Offers enhanced selectivity for aromatic compounds.