Learn More
Waters Corp Nova-Pak Silica Column, 60Å, 4 µm, 2.1 mm X 150 mm, 1/pk
Catalog No. 50785666 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Nova-Pak Silica columns are general purpose, normal-phase columns that are based on 4 µm particle technology. Compared to a 5 µm column, Nova-Pak columns offer higher resolution, more efficient separations, and faster chromatography.
Catalog No. 50-785-666 Supplier Waters Corp Supplier No. WAT052745
Nova-Pak Silica columns are general purpose, normal-phase columns that are based on 4 µm particle technology. Compared to a 5 µm column, Nova-Pak columns offer higher resolution, more efficient separations, and faster chromatography.