Learn More
Ihc World LLC NOVA ULTRA SAFRANIN STAIN KIT
SDP
Catalog No. NC0528301
Encompass
This kit is used for the detection of cartilage, mucin, and mast cell granules on formalin-fixed, paraffin-embedded tissue sections, and may be used for frozen sections as well.
Catalog No. NC0528301 Supplier Ihc World LLC Supplier No. IW3011A
This kit is used for the detection of cartilage, mucin, and mast cell granules on formalin-fixed, paraffin-embedded tissue sections, and may be used for frozen sections as well.