Learn More
Nova Biomedical Corporation Nova Vet™ Ketone/Glucose Meter – #53574
Catalog No. NC2882358
Encompass
Measures blood beta-hydroxybutyrate (BHB), the gold standard for detecting subclinical ketosisReady-to-use, disposable ketone test stripsNo calibration requiredSimple test procedure in the fieldAccurate, quantitative blood ketone results in 10 secondsGlucose testing can be performed using a separate glucose stripEarly identification and monitoring of changes in transition cow performanceProtection of herd milk production and qualityQuantification of herd prevalence of ketosisAvoidance of treatment costs for clinical ketosisMonitoring and management of herd feeding programProtection of herd reproductive performanceReduction in herd culling rates*Not for human use
Catalog No. NC2882358 Supplier Nova Biomedical Corporation Supplier No. 53574
Measures blood beta-hydroxybutyrate (BHB), the gold standard for detecting subclinical ketosisReady-to-use, disposable ketone test stripsNo calibration requiredSimple test procedure in the fieldAccurate, quantitative blood ketone results in 10 secondsGlucose testing can be performed using a separate glucose stripEarly identification and monitoring of changes in transition cow performanceProtection of herd milk production and qualityQuantification of herd prevalence of ketosisAvoidance of treatment costs for clinical ketosisMonitoring and management of herd feeding programProtection of herd reproductive performanceReduction in herd culling rates*Not for human use