Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novabiochem™ Capping Reagent A (20% n-Methylimidazole in Acetonitrile)

Catalog No. MBI02240505 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBI02240505 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBI02240505 Supplier MilliporeSigma™ Supplier No. US1BI0224/0505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in biosynthesis.

Applications:
  • DNA/ RNA synthesis with high yields
  • CAS: Acetonitrile: 75-05-8
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.