Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NovaBiochem™ Deblock Reagent, 10% DCA in Toluene

Catalog No. mbi0841np20 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBI0841NP20 20 L
MBI08414000 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MBI0841NP20 Supplier MilliporeSigma™ Supplier No. BI0841NP20
Only null left
Add to Cart
Add to Cart

Used for synthesis of DNA

  • Used in labeling, biosynthesis of DNA
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Concentration 10% (v/v)
Packaging Type Plastic barrel
Product Type DNA labeling reagents
Quantity 20 L
Synonym DCA in Toluene
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.