Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novabiochem™ Oxidizing Reagent, 0.05M, I2 in Pyridine

Catalog No. MBI04241005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBI04241005 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBI04241005 Supplier MilliporeSigma™ Supplier No. BI04241005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in biosynthesis.

  • % Pyridine concentration: 89.0 to 91.0%
  • Color: Dark brown
  • Component: Pyridine Purity: 99.0% min
  • Form: Liquid
  • Iodine Molarity: 0.04 to 0.06M
  • Water (H2O): 9.0 to 11.0%
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2.5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.